Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026374.3 | NP_080650.1 | 6 | 409 | ||
GenBank mRNA | AK012364.1 | - | 6 | 396 | ||
AK013858.1 | - | 6 | 455 | |||
AK031892.1 | - | 6 | 464 | |||
AK044885.1 | - | 6 | 455 | |||
AK078003.1 | - | 6 | 396 | |||
AK135539.1 | - | 6 | 396 | |||
AK157129.1 | - | 6 | 464 | |||
BC024718.1 | - | 6 | 409 | 5012365 |