Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026376.3 | NP_080652.2 | 5, 6 | 2066 | ||
GenBank mRNA | AK129175.1 | - | 5, 6 | 1331 | ||
AK147513.1 | - | 5, 6 | 2067 | |||
AK155833.1 | - | 3, 4 | 289 | |||
AY688678.1 | - | 5, 6 | 2026 |