Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pllp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026385.4 | NP_080661.1 | 4 | 523 | ||
GenBank mRNA | AK002477.1 | - | 4 | 524 | ||
AK018198.1 | - | 4 | 512 | |||
AK032540.1 | - | 4 | 512 | |||
BC024534.1 | - | 4 | 514 | 4987882 |