Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026396.3 | NP_080672.3 | 2 | 219 | ||
GenBank mRNA | AK002985.1 | - | 2 | 106 | ||
AK088313.1 | - | 2 | 203 | |||
AK168325.1 | - | 2 | 192 | |||
DT927497.1 | - | 2 | 219 |