Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026396.3 | NP_080672.3 | 9, 10 | 844 | ||
GenBank mRNA | AK010171.1 | - | 1, 2 | 62 | ||
AK088313.1 | - | 9, 10 | 832 | |||
AK168325.1 | - | 9, 10 | 817 | |||
BC014832.1 | - | 9, 10 | 814 | 4484024 | ||
CF749026.1 | - | 2, 3 | 246 |