Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026399.2 | NP_080675.2 | 2 | 268 | ||
GenBank mRNA | AK005790.1 | - | 2 | 208 | ||
AK020654.1 | - | 2 | 229 | |||
AY365434.1 | - | 2 | 268 | |||
BC005587.1 | - | 2 | 225 | |||
BC019369.1 | - | 2 | 191 | 4014928 | ||
BC020047.1 | - | 828 | ||||
BC034842.1 | - | 2 | 154 | |||
BC094624.1 | - | 2 | 205 |