Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026399.2 | NP_080675.2 | 4 | 478 | ||
GenBank mRNA | AK005790.1 | - | 4 | 418 | ||
AK020654.1 | - | 4 | 439 | |||
AY365434.1 | - | 4 | 478 | |||
BC005587.1 | - | 3 | 717 | |||
BC019369.1 | - | 4 | 401 | 4014928 | ||
BC020047.1 | - | 1320 | ||||
BC034842.1 | - | 3 | 646 |