Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190804.1 | NP_001177733.1 | 4 | 597 | ||
NM_001190805.1 | NP_001177734.1 | 4 | 597 | |||
NM_026400.5 | NP_080676.3 | 4 | 597 | |||
GenBank mRNA | AK010861.1 | - | 4 | 569 | ||
AK050500.1 | - | 4 | 556 | |||
AK075785.1 | - | 4 | 579 | |||
AK075953.1 | - | 4 | 559 | |||
AK148511.1 | - | 4 | 577 | |||
AK166730.1 | - | 4 | 555 | |||
AY054981.1 | - | 4 | 473 | |||
BC003999.2 | - | 4 | 528 | 3492402 | ||
BC018282.1 | - | 4 | 524 | 4207201 | ||
BC040747.1 | - | 4 | 528 | 3492402 |