Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026401.2 | NP_080677.1 | 2 | 346 | ||
GenBank mRNA | AB049958.1 | - | 2 | 307 | ||
AK003130.1 | - | 2 | 342 | |||
AK010282.1 | - | 2 | 351 | |||
AK011558.1 | - | 2 | 347 | |||
BC049555.1 | - | 341 | 6772707 | |||
BC055723.1 | - | 2 | 287 | 6411308 |