Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199151.1 | NP_001186080.1 | 10 | 2608 | ||
NM_001199153.1 | NP_001186082.1 | 10 | 2513 | |||
NM_001199154.1 | NP_001186083.1 | 10 | 2783 | |||
NM_026408.4 | NP_080684.3 | 10 | 2608 | |||
GenBank mRNA | AK013040.1 | - | 8 | 2342 | ||
AK017012.1 | - | 7 | 1543 | |||
AK081942.1 | - | 10 | 2787 | |||
AK082079.1 | - | 10 | 2612 | |||
AK132346.1 | - | 10 | 2711 | |||
AK160111.1 | - | 10 | 2612 | |||
BC098489.1 | - | 10 | 2512 | 30096466 | ||
BC130225.1 | - | 10 | 2498 | |||
BC152544.1 | - | 10 | 2496 |