Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190795.1 | NP_001177724.1 | 14 | 1720 | ||
NM_026409.4 | NP_080685.2 | 14 | 1732 | |||
GenBank mRNA | AK033741.1 | - | 14 | 1709 | ||
AK077525.1 | - | 14 | 1712 | |||
AK129402.1 | - | 14 | 1710 | |||
AK154422.1 | - | 14 | 1733 | |||
BC025218.1 | - | 4 | 565 | |||
BC043052.1 | - | 14 | 1670 | |||
BC117786.1 | - | 14 | 1694 | 40092124 | ||
BC117787.1 | - | 14 | 1694 | 40092125 |