Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024209.2 | NP_077171.1 | 7 | 825 | ||
GenBank mRNA | AK002764.1 | - | 7 | 787 | ||
AK008835.1 | - | 1 | 71 | |||
AK009104.1 | - | 7 | 805 | |||
BC002223.1 | - | 7 | 850 | 3491111 |