Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026416.2 | NP_080692.1 | 2 | 256 | ||
GenBank mRNA | AK009762.1 | - | 2 | 207 | ||
AK160178.1 | - | 2 | 207 | |||
AK160512.1 | - | 2 | 199 | |||
AK167006.1 | - | 2 | 199 | |||
AY221962.1 | - | 1 | 147 | |||
BC020031.1 | - | 2 | 256 | 3980274 |