Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021615.4 | NP_067628.1 | 3 | 1833 | ||
XM_005255955.4 | XP_005256012.1 | 4 | 2380 | |||
XM_011523085.2 | XP_011521387.1 | 3 | 2448 | |||
GenBank mRNA | AF280086.1 | - | 3 | 1841 |