Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024211.3 | NP_077173.1 | 5 | 783 | ||
GenBank mRNA | AK003957.1 | - | 4 | 422 | ||
AK009487.1 | - | 5 | 729 | |||
AK009824.1 | - | 5 | 683 | |||
AK168951.1 | - | 5 | 708 | |||
BC003455.1 | - | 5 | 759 | 2648336 | ||
BC019631.1 | - | 5 | 737 | 4023179 |