Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026417.4 | NP_080693.2 | 3 | 539 | ||
GenBank mRNA | AK009618.1 | - | 3 | 525 | ||
AK017721.1 | - | 3 | 517 | |||
AK017722.1 | - | 3 | 517 | |||
AK083301.1 | - | 3 | 519 | |||
AK159227.1 | - | 3 | 500 | |||
AK166347.1 | - | 3 | 527 | |||
BC003936.1 | - | 4 | 714 | 3486503 |