Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025091.1 | NP_001020262.1 | 5, 6 | 478 | ||
NM_001090.2 | NP_001081.1 | 5, 6 | 478 | |||
GenBank mRNA | AF027302.1 | - | 5, 6 | 477 | ||
AK298061.1 | - | 5, 6 | 421 | |||
AK307551.1 | - | 5, 6 | 432 | |||
BC016772.1 | - | 5, 6 | 415 | |||
BC034488.2 | - | 5, 6 | 423 | 4826422 | ||
BC112923.1 | - | 5, 6 | 411 | 7470309 |