Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026423.4 | NP_080699.3 | 3 | 654 | ||
GenBank mRNA | AK007709.1 | - | 3 | 615 | ||
AK010538.1 | - | 3 | 626 | |||
AK013882.1 | - | 3 | 623 | |||
AK047287.1 | - | 3 | 657 | |||
AK144807.1 | - | 3 | 595 | |||
AK164894.1 | - | 3 | 623 | |||
AK180146.1 | - | 2 | 146 | |||
BC025049.1 | - | 3 | 621 | 4951045 |