Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039710.1 | NP_001034799.1 | 4 | 642 | ||
NM_026424.3 | NP_080700.3 | 3 | 492 | |||
GenBank mRNA | AK002294.1 | - | 4 | 543 | ||
AK006551.1 | - | 3 | 493 | |||
AK012117.1 | - | 4 | 610 | |||
AK015454.1 | - | 4 | 642 | |||
AK136258.1 | - | 4 | 542 | |||
AK146260.1 | - | 4 | 536 | |||
AK168293.1 | - | 4 | 536 | |||
AK168837.1 | - | 4 | 540 | |||
BC062129.1 | - | 4 | 538 | 6506502 |