Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285452.1 | NP_001272381.1 | 3, 4 | 758 | ||
NM_028975.4 | NP_083251.2 | 4, 5 | 601 | |||
NM_030108.2 | NP_084384.1 | 4, 5 | 601 | |||
GenBank mRNA | AK005493.1 | - | 4, 5 | 493 | ||
AK007607.1 | - | 4, 5 | 409 | |||
AK007750.1 | - | 4, 5 | 490 | |||
AK017281.1 | - | 4, 5 | 470 | |||
AK019127.1 | - | 3, 4 | 427 | |||
AK032555.1 | - | 4, 5 | 508 | |||
AK153441.1 | - | 4, 5 | 426 | |||
AK165369.1 | - | 4, 5 | 403 | |||
AK206266.1 | - | 2, 3 | 221 | |||
BC005562.1 | - | 4, 5 | 402 | 3965864 | ||
BC016570.1 | - | 3, 4 | 474 | 3583937 | ||
BC023966.1 | - | 3, 4 | 467 | 5342147 |