Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026430.3 | NP_080706.1 | 6, 7 | 451 | ||
GenBank mRNA | AF399958.1 | - | 6, 7 | 451 | ||
AK005536.1 | - | 6, 7 | 391 | |||
AK075806.1 | - | 6, 7 | 503 | |||
AK076166.1 | - | 5, 6 | 492 | |||
AK152376.1 | - | 6, 7 | 506 | |||
AK208821.1 | - | 1, 2 | 190 | |||
BC037049.1 | - | 6, 7 | 497 | 4935237 |