Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197320.1 | NP_001184249.1 | 4 | 714 | ||
NM_021960.4 | NP_068779.1 | 3 | 1173 | |||
NM_182763.2 | NP_877495.1 | 2 | 925 | |||
GenBank mRNA | AF118124.1 | - | 3 | 1014 | ||
AF203373.1 | - | 2 | 717 | |||
AK294462.1 | - | 4 | 745 | |||
AK297217.1 | - | 4 | 709 | |||
AK300499.1 | - | 4 | 819 | |||
AK304775.1 | - | 4 | 855 | |||
AK312508.1 | - | 3 | 1045 | |||
AK316267.1 | - | 4 | 1001 | |||
BC017197.2 | - | 3 | 1019 | 3138465 | ||
BC071897.1 | - | 3 | 1021 | 6586159 | ||
BC107735.1 | - | 3 | 1036 | 4770487 | ||
BT006640.1 | - | 3 | 965 | |||
L08246.1 | - | 3 | 1025 |