Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197320.1 | NP_001184249.1 | 4 | 1591 | ||
NM_021960.4 | NP_068779.1 | 3 | 2050 | |||
NM_182763.2 | NP_877495.1 | 2 | 1802 | |||
GenBank mRNA | AF118124.1 | - | 3 | 1893 | ||
BC017197.2 | - | 3 | 1896 | 3138465 | ||
BC071897.1 | - | 3 | 1898 | 6586159 | ||
L08246.1 | - | 3 | 1904 |