Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167829.1 | NP_001161301.1 | 2 | 176 | ||
NM_026437.3 | NP_080713.2 | 2 | 176 | |||
GenBank mRNA | AK004643.1 | - | 2 | 176 | ||
AK007867.1 | - | 2 | 136 | |||
AK145769.1 | - | 2 | 142 | |||
AK168474.1 | - | 2 | 170 | |||
AK171767.1 | - | 2 | 176 | |||
AK202624.1 | - | 2 | 140 | |||
BC013514.1 | - | 2 | 135 | 4235293 | ||
BY122265.1 | - | 2 | 176 |