Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170953.1 | NP_001164424.1 | 5, 6 | 869 | ||
NM_026440.4 | NP_080716.1 | 5, 6 | 869 | |||
GenBank mRNA | AK011300.1 | - | 5, 6 | 853 | ||
AK015403.1 | - | 5, 6 | 826 | |||
AK031780.1 | - | 5, 6 | 827 | |||
AK082331.1 | - | 5, 6 | 836 | |||
AK129130.1 | - | 4, 5 | 806 | |||
BC021794.1 | - | 5, 6 | 815 | 4208274 |