Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170953.1 | NP_001164424.1 | 4 | 653 | ||
NM_026440.4 | NP_080716.1 | 4 | 653 | |||
GenBank mRNA | AK011300.1 | - | 4 | 637 | ||
AK015403.1 | - | 4 | 610 | |||
AK031780.1 | - | 4 | 611 | |||
AK082331.1 | - | 4 | 620 | |||
AK129130.1 | - | 3 | 590 | |||
BC021794.1 | - | 4 | 599 | 4208274 |