Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026440.4 | NP_080716.1 | 7 | 1205 | ||
GenBank mRNA | AK011300.1 | - | 7 | 1189 | ||
AK031780.1 | - | 7 | 1163 | |||
AK082331.1 | - | 7 | 1172 | |||
AK129130.1 | - | 6 | 1142 | |||
BC021794.1 | - | 7 | 1151 | 4208274 |