Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026449.3 | NP_080725.2 | 6 | 1033 | ||
GenBank mRNA | AK005605.1 | - | 6 | 1054 | ||
AK006193.1 | - | 6 | 1012 | |||
AK016415.1 | - | 6 | 1034 | |||
BC049554.1 | - | 6 | 1015 | |||
BC100447.1 | - | 6 | 886 | |||
BC145757.1 | - | 6 | 893 | 40131060 |