Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026452.2 | NP_080728.1 | 7 | 825 | ||
GenBank mRNA | AK004527.1 | - | 3 | 250 | ||
AK009179.1 | - | 7 | 837 | |||
AK154343.1 | - | 7 | 850 | |||
AK158979.1 | - | 7 | 844 | |||
BC036386.1 | - | 7 | 825 | 1067021 |