Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mak16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026453.3 | NP_080729.1 | 7, 8 | 559 | ||
GenBank mRNA | AK015369.1 | - | 7, 8 | 600 | ||
AK075982.1 | - | 7, 8 | 560 | |||
AK075983.1 | - | 7, 8 | 560 | |||
BC013079.1 | - | 7, 8 | 574 | 3499754 |