Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026454.3 | NP_080730.1 | 7 | 533 | ||
GenBank mRNA | AK007549.1 | - | 7 | 504 | ||
AK007937.1 | - | 7 | 533 | |||
AK007960.1 | - | 7 | 523 | |||
AK010957.1 | - | 8 | 610 | |||
AK030594.1 | - | 513 | ||||
AK153481.1 | - | 7 | 534 | |||
AK208168.1 | - | 6 | 317 | |||
BC006057.1 | - | 1 | 46 | |||
BC016117.1 | - | 8 | 815 | 4486610 |