Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpini2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026460.3 | NP_080736.2 | 8, 9 | 1194 | ||
GenBank mRNA | AF251276.1 | - | 8, 9 | 1179 | ||
AK007347.1 | - | 8, 9 | 1195 | |||
AK007510.1 | - | 8, 9 | 1169 | |||
BC098466.1 | - | 8, 9 | 1171 | 6433250 |