Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023402.2 | NP_075891.1 | 3 | 261 | ||
GenBank mRNA | AK002885.1 | - | 3 | 263 | ||
AK005133.1 | - | 3 | 303 | |||
AK146115.1 | - | 3 | 279 | |||
AK160123.1 | - | 3 | 303 | |||
AK168316.1 | - | 3 | 262 | |||
BC028878.1 | - | 3 | 243 | 4218748 | ||
BC099425.1 | - | 2 | 232 | 30847912 | ||
BC131925.1 | - | 3 | 237 | 40130202 | ||
BC131927.1 | - | 3 | 237 | 40130204 |