Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prorsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026465.3 | NP_080741.1 | 1, 2 | 121 | ||
GenBank mRNA | AK008574.1 | - | 1, 2 | 122 | ||
AK019442.1 | - | 1, 2 | 101 | |||
BC115900.1 | - | 1, 2 | 122 | 40091622 |