Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006739.3 | NP_006730.2 | 12 | 1649 | ||
XM_006724242.3 | XP_006724305.1 | 12 | 1704 | |||
GenBank mRNA | AB209612.1 | - | 6 | 1137 | ||
AK122853.1 | - | 11 | 1497 | |||
AK130620.1 | - | 11 | 1603 | |||
AK223323.1 | - | 12 | 1639 | |||
AK291136.1 | - | 12 | 1630 | |||
AK298624.1 | - | 12 | 1526 | |||
AK301121.1 | - | 567 | ||||
AK308343.1 | - | 10 | 1250 | |||
AK312533.1 | - | 12 | 1629 | |||
BC000142.1 | - | 12 | 1601 | 2900229 | ||
BC003656.2 | - | 11 | 1575 | 3542201 | ||
CR456517.1 | - | 12 | 1589 | |||
D83986.1 | - | 12 | 1637 | |||
X74795.2 | - | 11 | 1577 |