Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175127.2 | NP_780336.1 | 5 | 796 | ||
GenBank mRNA | AK076211.1 | - | 5 | 802 | ||
AK077653.1 | - | 5 | 809 | |||
AK122295.1 | - | 5 | 781 | |||
AK162188.1 | - | 5 | 677 | |||
BC055458.1 | - | 4 | 498 | |||
BC078447.1 | - | 2 | 644 | |||
BC094542.1 | - | 5 | 796 | 6408744 |