Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164600.1 | NP_001158072.1 | 7 | 684 | ||
NM_028364.2 | NP_082640.2 | 8 | 765 | |||
NM_133691.5 | NP_598452.2 | 7 | 714 | |||
GenBank mRNA | AK012434.1 | - | 7 | 654 | ||
AK075960.1 | - | 7 | 682 | |||
AK146534.1 | - | 8 | 736 | |||
AK149632.1 | - | 8 | 731 | |||
AK151526.1 | - | 7 | 678 | |||
AK168789.1 | - | 7 | 681 | |||
AK181151.1 | - | 3 | 291 | |||
AK181470.1 | - | 3 | 291 | |||
AK189419.1 | - | 3 | 290 | |||
BC010601.1 | - | 7 | 684 | 3257153 | ||
BC119437.1 | - | 5 | 421 | |||
BC148274.1 | - | 5 | 421 |