Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025091.1 | NP_001020262.1 | 7, 8 | 661 | ||
NM_001090.2 | NP_001081.1 | 7, 8 | 661 | |||
GenBank mRNA | AF027302.1 | - | 7, 8 | 660 | ||
AK298061.1 | - | 7, 8 | 604 | |||
AK307551.1 | - | 7, 8 | 615 | |||
AL832430.1 | - | 1, 2 | 91 | |||
BC016772.1 | - | 7, 8 | 598 | |||
BC034488.2 | - | 7, 8 | 606 | 4826422 | ||
BC112923.1 | - | 7, 8 | 594 | 7470309 | ||
BI547952.1 | - | 7, 8 | 683 |