Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133692.2 | NP_598453.1 | 4 | 318 | ||
GenBank mRNA | AF294329.1 | - | 4 | 329 | ||
AK010805.1 | - | 4 | 314 | |||
AK031778.1 | - | 4 | 320 | |||
AK088385.1 | - | 4 | 311 | |||
AK168179.1 | - | 4 | 306 | |||
AK180448.1 | - | 4 | 238 | |||
BC055325.1 | - | 4 | 321 | 6402059 |