Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133692.2 | NP_598453.1 | 5 | 436 | ||
GenBank mRNA | AF294329.1 | - | 5 | 447 | ||
AK010805.1 | - | 5 | 432 | |||
AK031778.1 | - | 5 | 438 | |||
AK088385.1 | - | 5 | 429 | |||
AK168179.1 | - | 5 | 424 | |||
AK180448.1 | - | 5 | 356 | |||
AK189114.1 | - | 5 | 357 | |||
BC055325.1 | - | 5 | 439 | 6402059 |