Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ooep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026480.3 | NP_080756.1 | 3 | 465 | ||
GenBank mRNA | AB283026.1 | - | 3 | 405 | ||
AK010266.1 | - | 3 | 460 | |||
AK010809.1 | - | 3 | 466 | |||
AK133554.1 | - | 3 | 430 | |||
AK135950.1 | - | 3 | 431 | |||
AK136199.1 | - | 431 | ||||
AK145510.1 | - | 3 | 431 | |||
BC062245.1 | - | 3 | 416 | 3470921 | ||
DQ238107.1 | - | 3 | 466 |