Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tppp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026481.3 | NP_080757.1 | 3 | 354 | ||
GenBank mRNA | AK009771.1 | - | 3 | 318 | ||
AK012437.1 | - | 3 | 322 | |||
AK155018.1 | - | 3 | 325 | |||
AK160507.1 | - | 3 | 325 | |||
BC010788.1 | - | 3 | 311 | 4216285 | ||
BC116622.1 | - | 2 | 146 | |||
BF469439.1 | - | 3 | 354 |