Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026483.2 | NP_080759.2 | 4 | 1103 | ||
GenBank mRNA | AK013337.1 | - | 4 | 1091 | ||
BC018177.1 | - | 4 | 1097 | |||
BC049270.1 | - | 4 | 1079 | 3988645 | ||
CK390971.1 | - | 3 | 502 |