Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hormad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289532.1 | NP_001276461.1 | 9 | 533 | ||
NM_001289534.1 | NP_001276463.1 | 9 | 533 | |||
NM_001289537.1 | NP_001276466.1 | 9 | 533 | |||
NM_026489.3 | NP_080765.1 | 9 | 524 | |||
GenBank mRNA | AK014945.1 | - | 9 | 532 | ||
AK016324.1 | - | 9 | 532 | |||
AK016743.1 | - | 9 | 527 | |||
AK163995.1 | - | 9 | 532 | |||
AY626343.1 | - | 9 | 532 | |||
AY634284.1 | - | 9 | 532 | |||
BC051129.1 | - | 9 | 532 | 6706580 |