Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026500.3 | NP_080776.1 | 2 | 505 | ||
GenBank mRNA | AK004833.1 | - | 2 | 506 | ||
AK013179.1 | - | 2 | 467 | |||
AK076462.1 | - | 2 | 460 | |||
AK133731.1 | - | 2 | 467 | |||
BC023840.1 | - | 2 | 432 | 5344890 |