Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195634.2 | NP_001182563.2 | 2 | 271 | ||
GenBank mRNA | AK008377.1 | - | 2 | 272 | ||
AK008459.1 | - | 2 | 274 |
Gene Symbol: Defa20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183268.4 | NP_899091.2 | 2 | 271 | ||
GenBank mRNA | AK008107.1 | - | 2 | 272 | ||
AK008275.1 | - | 2 | 272 | |||
DQ103703.1 | - | 2 | 228 |
Gene Symbol: Gm15308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177521.1 | NP_001170992.1 | 273 |
Gene Symbol: Gm21002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270555.1 | NP_001257484.1 | 273 | |||
GenBank mRNA | AY746425.1 | - | 228 |