Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bambi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026505.2 | NP_080781.1 | 2 | 561 | ||
GenBank mRNA | AF153440.1 | - | 2 | 547 | ||
AK011305.1 | - | 2 | 584 | |||
AK017303.1 | - | 2 | 563 | |||
AK045192.1 | - | 2 | 561 | |||
AK143185.1 | - | 2 | 576 | |||
AK164632.1 | - | 2 | 564 | |||
AK212745.1 | - | 1 | 114 | |||
BC019378.1 | - | 2 | 543 | 5009872 |