Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026506.2 | NP_080782.1 | 1, 2 | 84 | ||
GenBank mRNA | AK011129.1 | - | 1, 2 | 83 | ||
AK012802.1 | - | 1, 2 | 73 | |||
AK013235.1 | - | 1, 2 | 85 | |||
BC027499.1 | - | 1, 2 | 57 | 1430048 | ||
BC051470.1 | - | 1, 2 | 129 | 1244608 | ||
BC094411.1 | - | 1, 2 | 71 | 1380524 |