Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwilch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026507.4 | NP_080783.3 | 13 | 1326 | ||
GenBank mRNA | AK009559.1 | - | 13 | 1297 | ||
AK019825.1 | - | 13 | 1304 | |||
AK050917.1 | - | 13 | 1292 | |||
BC027435.1 | - | 13 | 1274 | 2812472 |