Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apopt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026511.2 | NP_080787.1 | 4 | 494 | ||
GenBank mRNA | AK002873.1 | - | 4 | 496 | ||
AK005403.1 | - | 4 | 482 | |||
AK010493.1 | - | 4 | 484 | |||
AK012648.1 | - | 4 | 492 | |||
AK029408.1 | - | 4 | 470 | |||
BC071262.1 | - | 4 | 494 | 6509633 |